COM638471461 Combi-Blocks, Inc.

1-(3-Chloro-4-methylphenyl)urea

  • MDL number MFCD00025419
  • Cas Number : 13142-64-8
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8384
Availability details for
Quantity
  • Availability
  • Add to Cart