COM638471468 Combi-Blocks, Inc.

4-Phenyl-3-(1h-pyrrol-1-yl)thiophene-2-carbohydrazide

  • MDL number MFCD26130164
  • Cas Number : 1779132-51-2
  • Molecular Weight : 283.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8424
Availability details for
Quantity
  • Availability
  • Add to Cart