COM638471490 Combi-Blocks, Inc.

2-(2-Phenylhydrazinyl)pyridine-3-carboxylic acid

  • MDL number MFCD19705704
  • Cas Number : 1325304-37-7
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8536

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8536
Availability details for
Quantity
  • Availability
  • Add to Cart