COM638471518 Combi-Blocks, Inc.

7-(4-Chlorophenyl)-2-(methylthio)thieno[3,2-d]pyrimidin-4(3h)-one

  • MDL number MFCD26130183
  • Cas Number : 1779132-84-1
  • Molecular Weight : 308.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8703
Availability details for
Quantity
  • Availability
  • Add to Cart