COM638471522 Combi-Blocks, Inc.

4-Chloro-2-((phenylamino)methyl)phenol

  • MDL number MFCD10699477
  • Cas Number : 7123-49-1
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8727
Availability details for
Quantity
  • Availability
  • Add to Cart