COM638471530 Combi-Blocks, Inc.

5-Ethyl-1-(2-fluorobenzyl)-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD17023509
  • Cas Number : 1267882-46-1
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8762
Availability details for
Quantity
  • Availability
  • Add to Cart