COM638471536 Combi-Blocks, Inc.

(4R,12As)-7-methoxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2h-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxylic acid

  • MDL number MFCD28978321
  • Cas Number : 1335210-34-8
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8792
Availability details for
Quantity
  • Availability
  • Add to Cart