COM638471550 Combi-Blocks, Inc.

2-(4-Fluorophenyl)-2-oxoethyl 2-(((benzyloxy)carbonyl)amino)-2-methylpropanoate

  • MDL number MFCD29921977
  • Cas Number : 1261118-01-7
  • Molecular Weight : 373.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8857
Availability details for
Quantity
  • Availability
  • Add to Cart