COM638471552 Combi-Blocks, Inc.

4-([3-(4-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methyl)piperidine

  • MDL number MFCD16874868
  • Cas Number : 1239771-50-6
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8862
Availability details for
Quantity
  • Availability
  • Add to Cart