COM638471555 Combi-Blocks, Inc.

3'-O-Methylguanosine

  • MDL number MFCD00057057
  • Cas Number : 10300-27-3
  • Molecular Weight : 297.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8889
Availability details for
Quantity
  • Availability
  • Add to Cart