COM638471557 Combi-Blocks, Inc.

4A,5,6,7,8,9-Hexahydrothieno[3',2':4,5]pyrimido[1,2-a]azepin-11(4h)-one

  • MDL number MFCD26130227
  • Cas Number : 1707737-26-5
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8904
Availability details for
Quantity
  • Availability
  • Add to Cart