COM638471566 Combi-Blocks, Inc.

1-([(2E)-3-Phenylprop-2-enoyl]oxy)pyrrolidine-2,5-dione

  • MDL number MFCD00435763
  • Cas Number : 23776-87-6
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-8971
Availability details for
Quantity
  • Availability
  • Add to Cart