COM638471574 Combi-Blocks, Inc.

1-(4-Fluoro-3-methylphenyl)-5-methyl-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD14703541
  • Cas Number : 1239728-42-7
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9032
Availability details for
Quantity
  • Availability
  • Add to Cart