COM638471643 Combi-Blocks, Inc.

1-([3-(4-Ethoxyphenyl)-1h-pyrazol-5-yl]carbonyl)piperidine

  • MDL number MFCD14920187
  • Cas Number : 1093657-24-9
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9401

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9401
Availability details for
Quantity
  • Availability
  • Add to Cart