COM638471648 Combi-Blocks, Inc.

Ethyl 2-methyl-2h-1,2,6-thiadiazine-4-carboxylate 1,1-dioxide

  • MDL number MFCD26130284
  • Cas Number : 1710202-97-3
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9416
Availability details for
Quantity
  • Availability
  • Add to Cart