COM638471650 Combi-Blocks, Inc.

2-Chloro-n-methylpyridine-3-sulfonamide

  • MDL number MFCD01860228
  • Cas Number : 38030-51-2
  • Molecular Weight : 206.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9422
Availability details for
Quantity
  • Availability
  • Add to Cart