COM638471669 Combi-Blocks, Inc.

6-Chloro-9-(2,3,5-tri-o-benzoyl-2-c-methyl-beta-d-ribofuranosyl)-9h-purine

  • MDL number MFCD28957715
  • Cas Number : 205171-04-6
  • Molecular Weight : 613.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9554
Availability details for
Quantity
  • Availability
  • Add to Cart