COM638471680 Combi-Blocks, Inc.

1-[3-(Benzoyloxy)propyl]-2,3-dihydro-5-(2-nitropropyl)-1h-indole-7-carbonitrile

  • MDL number MFCD17169997
  • Cas Number : 350797-56-7
  • Molecular Weight : 393.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9627

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9627
Availability details for
Quantity
  • Availability
  • Add to Cart