COM638471690 Combi-Blocks, Inc.

5-[2-(1-tert-Butoxycarbonyl-piperidin-4-yl)-ethyl]-4-oxo-3a,4,5,6,7,8-hexahydro-3h-1,5,8a-triaza-azulene-2-carboxylic acid

  • MDL number MFCD29059076
  • Cas Number : 163213-39-6
  • Molecular Weight : 408.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9684
Availability details for
Quantity
  • Availability
  • Add to Cart