COM638471694 Combi-Blocks, Inc.

Cbz-l-phenylalaninal

  • MDL number MFCD00798796
  • Cas Number : 59830-60-3
  • Molecular Weight : 283.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9705
Availability details for
Quantity
  • Availability
  • Add to Cart