COM638471695 Combi-Blocks, Inc.

4-Chloro-2-[(4-methoxyanilino)methyl]benzenol

  • MDL number MFCD05154999
  • Cas Number : 98591-11-8
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9707
Availability details for
Quantity
  • Availability
  • Add to Cart