COM638471702 Combi-Blocks, Inc.

2'-Deoxy-2'-iodouridine

  • MDL number MFCD28125329
  • Cas Number : 4753-03-1
  • Molecular Weight : 354.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9746
Availability details for
Quantity
  • Availability
  • Add to Cart