COM638471716 Combi-Blocks, Inc.

1-(2-Ethoxyphenyl)urea

  • MDL number MFCD00995172
  • Cas Number : 148-70-9
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9824

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9824
Availability details for
Quantity
  • Availability
  • Add to Cart