COM638471729 Combi-Blocks, Inc.

1-(2-Methoxybenzyl)guanidine

  • MDL number MFCD00607784
  • Cas Number : 224947-74-4
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9883
Availability details for
Quantity
  • Availability
  • Add to Cart