COM638471738 Combi-Blocks, Inc.

1-(4-Chlorophenyl)-5-(pyridin-3-yl)-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD14703474
  • Cas Number : 1030089-58-7
  • Molecular Weight : 300.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-9943
Availability details for
Quantity
  • Availability
  • Add to Cart