COM638471768 Combi-Blocks, Inc.

7-Fluoro-2-methanesulfonyl-3,4-dihydro-1H-isoquinoline

  • MDL number MFCD30527683
  • Cas Number : 935543-62-7
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2296

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2296
Availability details for
Quantity
  • Availability
  • Add to Cart