COM638471770 Combi-Blocks, Inc.

1,3-Dimethyl 2-[3-cyano-5-(trifluoromethyl)pyridin-2-yl]propanedioate

  • MDL number MFCD30527685
  • Cas Number : 2055119-00-9
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2302
Availability details for
Quantity
  • Availability
  • Add to Cart