COM638471779 Combi-Blocks, Inc.

2-(Dimethoxymethyl)pyridine-4-carbaldehyde

  • MDL number MFCD30527689
  • Cas Number : 1625618-11-2
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2325
Availability details for
Quantity
  • Availability
  • Add to Cart