COM638471789 Combi-Blocks, Inc.

N-[2-(4-Nitrophenyl)ethyl]methanesulfonamide

  • MDL number MFCD14879485
  • Cas Number : 24954-60-7
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6336
Availability details for
Quantity
  • Availability
  • Add to Cart