COM638471790 Combi-Blocks, Inc.

N-[2-(4-Aminophenyl)ethyl]methanesulfonamide

  • MDL number MFCD11936864
  • Cas Number : 24954-59-4
  • Molecular Weight : 214.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6337
Availability details for
Quantity
  • Availability
  • Add to Cart