COM638471794 Combi-Blocks, Inc.

1-(2-Iodoethyl)-2-pyrrolidinone

  • MDL number MFCD14583070
  • Cas Number : 713521-27-8
  • Molecular Weight : 239.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6360
Availability details for
Quantity
  • Availability
  • Add to Cart