COM638471798 Combi-Blocks, Inc.

2-Chloro-4-(imidazol-1-yl)benzonitrile

  • MDL number MFCD17256773
  • Cas Number : 1007318-66-2
  • Molecular Weight : 203.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6375
Availability details for
Quantity
  • Availability
  • Add to Cart