COM638471801 Combi-Blocks, Inc.

tert-Butyl 4-(hydroxymethyl)-4-(prop-2-en-1-yl)piperidine-1-carboxylate

  • MDL number MFCD17676692
  • Cas Number : 236406-37-4
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6384
Availability details for
Quantity
  • Availability
  • Add to Cart