COM638471803 Combi-Blocks, Inc.

3-(Dimethoxymethyl)-5-(methylsulfanyl)pyridine

  • MDL number MFCD30527692
  • Cas Number : 2055119-33-8
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6388
Availability details for
Quantity
  • Availability
  • Add to Cart