COM638471805 Combi-Blocks, Inc.

1-tert-Butyl 2-ethyl 2-(prop-2-en-1-yl)pyrrolidine-1,2-dicarboxylate

  • MDL number MFCD18909479
  • Cas Number : 1333319-68-8
  • Molecular Weight : 283.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6395
Availability details for
Quantity
  • Availability
  • Add to Cart