COM638471808 Combi-Blocks, Inc.

Methyl 5-(dimethoxymethyl)pyridine-3-carboxylate

  • MDL number MFCD30527693
  • Cas Number : 2055119-34-9
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6427
Availability details for
Quantity
  • Availability
  • Add to Cart