COM638471824 Combi-Blocks, Inc.

2,2-Dimethyl-propionic acid 6-methoxy-1-oxo-1,3-dihydro-isoindol-2-ylmethyl ester

  • MDL number MFCD09991673
  • Cas Number : 944718-09-6
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8193
Availability details for
Quantity
  • Availability
  • Add to Cart