COM638471847 Combi-Blocks, Inc.

4'-Acetoxy-biphenyl-4-carboxylic acid

  • MDL number MFCD00267980
  • Cas Number : 75175-09-6
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-0691
Availability details for
Quantity
  • Availability
  • Add to Cart