COM638471870 Combi-Blocks, Inc.

5-(2-Bromo-4-nitrophenyl)-2-furaldehyde

  • MDL number MFCD00297692
  • Cas Number : 327049-95-6
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2145
Availability details for
Quantity
  • Availability
  • Add to Cart