COM638471878 Combi-Blocks, Inc.

3-(3,4-Dihydroisoquinolin-2(1h)-yl)propan-1-amine

  • MDL number MFCD06446877
  • Cas Number : 5596-87-2
  • Molecular Weight : 190.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2249
Availability details for
Quantity
  • Availability
  • Add to Cart