COM638471880 Combi-Blocks, Inc.

1-(P-Tolyl)-3-adamantanecarboxylic acid

  • MDL number MFCD00267983
  • Cas Number : 56531-69-2
  • Molecular Weight : 270.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2276
Availability details for
Quantity
  • Availability
  • Add to Cart