COM638471881 Combi-Blocks, Inc.

4-([Cyclohexyl(methyl)amino]sulfonyl)benzoic acid

  • MDL number MFCD03116971
  • Cas Number : 101116-05-6
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2344
Availability details for
Quantity
  • Availability
  • Add to Cart