COM638471890 Combi-Blocks, Inc.

4-P-Tolylsulfamoyl-benzoic acid

  • MDL number MFCD03147233
  • Cas Number : 379254-26-9
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2831

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2831
Availability details for
Quantity
  • Availability
  • Add to Cart