COM638471892 Combi-Blocks, Inc.

[2-(2,6-Dichlorophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06340082
  • Cas Number : 757192-72-6
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-2901
Availability details for
Quantity
  • Availability
  • Add to Cart