COM638471925 Combi-Blocks, Inc.

3-(4-Methyl-piperazine-1-sulfonyl)-benzoic acid

  • MDL number MFCD02725037
  • Cas Number : 380339-63-9
  • Molecular Weight : 284.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4247
Availability details for
Quantity
  • Availability
  • Add to Cart