COM638471932 Combi-Blocks, Inc.

1-Benzenesulfonyl-pyrrolidine-2-carboxylic acid

  • MDL number MFCD00579687
  • Cas Number : 88425-47-2
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4434

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4434
Availability details for
Quantity
  • Availability
  • Add to Cart