COM638471964 Combi-Blocks, Inc.

2-[(Anilinocarbonyl)amino]-3-methylbutanoic acid

  • MDL number MFCD00483843
  • Cas Number : 84860-35-5
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6446
Availability details for
Quantity
  • Availability
  • Add to Cart