COM638471983 Combi-Blocks, Inc.

9-Chloro-2,3-dihydro-1h-cyclopenta[b]quinoline

  • MDL number MFCD05670710
  • Cas Number : 40528-00-5
  • Molecular Weight : 203.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6972
Availability details for
Quantity
  • Availability
  • Add to Cart