COM638472028 Combi-Blocks, Inc.

1-((4-Chlorophenyl)methyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD03966892
  • Cas Number : 34569-29-4
  • Molecular Weight : 221.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9446
Availability details for
Quantity
  • Availability
  • Add to Cart