COM638472055 Combi-Blocks, Inc.

D-Phenylalanine methyl ester

  • MDL number MFCD07786601
  • Cas Number : 21685-51-8
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0083
Availability details for
Quantity
  • Availability
  • Add to Cart